Printer Ribbons, M710 R6600 Series
Supplier: Brady
173188
BRDY173188EA 65.22 GBP
BRDY173188
Printer Ribbons, M710 R6600 Series
Printer Ribbons
One-color ribbon for printing a single color onto a label.
  • R6600 series resin formulation offers excellent solvent and friction resistance when printed on the appropriate Brady® label material
  • Recommended for printing on heat shrinkable tubing materials from Brady®, including material numbers B-342, B-7641, B-7642, B-7646, and B-7647
  • Self-contained M710 series ribbon cartridges insert straight into the printer with ease
M710 Series snap-in print ribbons enable 5 second ribbon supply changes. No calibrating, no threading, no waste, no hassles, and theMore Product Information