Self-Laminating Aggressive Adhesive Vinyl Wrap Around Labels for Wraptor A6200 Printers
Supplier: Brady
152201
BRDY152201EA 36.44 GBP
BRDY152201
Self-Laminating Aggressive Adhesive Vinyl Wrap Around Labels for Wraptor A6200 Printers
Labels
Keep your wire identification secure with this self-laminating marker with extra-strong adhesive.
  • The extra-strong adhesive makes these markers ideal for small-gauge wires
  • Features a white printable area and a clear tail that wraps around the marker
  • Tail serves as an overlaminate to protect the print
  • A ribbon is required to print this material
Abrasion-Resistant, chemical-resistant, humidity-resistant, oil-resistant, water-resistant, weather-resistant.
Certifications: Compliant with Directive 2011/65/EU and 2015/863.