Thermal transfer printable labels, series R6400
Supplier: Brady
120999 121003 121004 121005 121009 121010 121011 121012
BRDY120999EA 141.92 GBP
BRDY120999 BRDY121003 BRDY121004 BRDY121005 BRDY121009 BRDY121010 BRDY121011 BRDY121012
Thermal transfer printable labels, series R6400
Labels
Polypropylene (B-7425) material is ideal for general purpose barcode applications and withstands liquid nitrogen, autoclave and other harsh environments.
  • Die-cut labels are on a roll
  • General identification, laboratory labelling and temperature indicating labelling applications
  • Service temperature: –80 to 70 °C
  • This R6400 series has matt finish
  • Thickness: 0,102 mm
  • Material with autoclavable and cryogenic storage properties
This node contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates.More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable