Anti-PP4R3B Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
BS-8328R-A680
BOSSBS-8328R-A680EA 837 GBP
BOSSBS-8328R-A680
Anti-PP4R3B Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
SMEK2 contains a WH1 domain. There are five named isoforms produced by alternative splicing. Mapping of a diet-induced hypercholesterolemia locus (Dihc2) in rats identified SMEK2 as a strong candidate for responsiveness to dietary cholesterol.

Type: Primary
Antigen: PP4R3B
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 701-800/849
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR