Anti-NIR2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-7763R-A350
BOSSBS-7763R-A350EA 837 GBP
BOSSBS-7763R-A350
Anti-NIR2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Regulates RHOA activity, and plays a role in cytoskeleton remodeling. Necessary for normal completion of cytokinesis. Plays a role in maintaining normal diacylglycerol levels in the Golgi apparatus. Binds phosphatidyl inositol phosphates (in vitro). May catalyze the transfer of phosphatidylinositol and phosphatidylcholine between membranes (By similarity). Necessary for maintaining the normal structure of the endoplasmic reticulum and the Golgi apparatus. Required for protein export from the endoplasmic reticulum and the Golgi. Binds calcium ions.

Recommended Dilutions: IF(IHC-P): 1:50-200More Product Information
Order Now

SPECIFICATIONS

Antigen symbol NIR2
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 350
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 9600
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR