Anti-SUMO2/3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: Bioss
BS-7338R-FITC
BOSSBS-7338R-FITCEA 843 GBP
BOSSBS-7338R-FITC
Anti-SUMO2/3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
The small ubiquitin-related modifier (SUMO) proteins, which include SUMO-1, SUMO-2 and SUMO-3, belong to the ubiquitin-like protein family. Like ubiquitin, the SUMO proteins are synthesized as precursor proteins that undergo processing before conjugation to target proteins. Also, both utilize the E1, E2, and E3 cascade enzymes for conjugation. However, SUMO and ubiquitin differ with respect to targeting. Ubiquitination predominantly targets proteins for degradation, whereas sumoylation targets proteins to a variety of cellular processing, including nuclear transport, transcriptionalMore Product Information