Anti-SEPT5 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-6905R-CY5
BOSSBS-6905R-CY5EA 843 GBP
BOSSBS-6905R-CY5
Anti-SEPT5 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
This gene is a member of the septin gene family of nucleotide binding proteins, originally described in yeast as cell division cycle regulatory proteins. Septins are highly conserved in yeast, Drosophila, and mouse and appear to regulate cytoskeletal organization. Disruption of septin function disturbs cytokinesis and results in large multinucleate or polyploid cells. This gene is mapped to 22q11, the region frequently deleted in DiGeorge and velocardiofacial syndromes. A translocation involving the MLL gene and this gene has also been reported in patients with acute myeloid leukemia.More Product Information
Order Now

SPECIFICATIONS

Antigen symbol SEPT5
Antigen name Septin 5
Environmentally Preferable
Antigen synonyms PNUTL1|CDCREL1|H5|HCDCREL-1|CDCREL|CDCREL-1
Antibody type Primary
Clonality Polyclonal
Conjugation Cy5®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 5413
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR