Anti-WDR16 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-6750R-CY5
BOSSBS-6750R-CY5EA 864 GBP
BOSSBS-6750R-CY5
Anti-WDR16 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
WD repeat containing proteins, such as WDR16, play crucial roles in a wide range of physiologic functions, including signal transduction, RNA processing, remodeling the cytoskeleton, regulation of vesicular traffic, and cell division.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: WDR16
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR