Anti-NTN4 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-6711R
BOSSBS-6711REA 679 GBP
BOSSBS-6711R
Anti-NTN4 Rabbit Polyclonal Antibody
Antibodies
May play an important role in neural, kidney and vascular development. Promotes neurite elongation from olfactory bulb explants.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: NTN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR