Anti-TLR7 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-6601R-CY7
BOSSBS-6601R-CY7EA 857 GBP
BOSSBS-6601R-CY7
Anti-TLR7 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Key component of innate and adaptive immunity. TLRs (Toll-like receptors) control host immune response against pathogens through recognition of molecular patterns specific to microorganisms. TLR7 is a nucleotide-sensing TLR which is activated by single-stranded RNA. Acts via MYD88 and TRAF6, leading to NF-kappa-B activation, cytokine secretion and the inflammatory response (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TLR7
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol TLR7
Environmentally Preferable
Antigen name Toll-like Receptor 7
Antigen synonyms TLR7|UNQ248|Toll-like receptor 7|Anti-TLR7|PRO285
Antibody type Primary
Clonality Polyclonal
Conjugation Cy7®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 51284
Isotype IgG
ImmunoChemistry Yes
Immunogen 500-550/1049
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR