Anti-APOC4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
BS-6338R-A680
BOSSBS-6338R-A680EA 837 GBP
BOSSBS-6338R-A680
Anti-APOC4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
This gene encodes a lipid-binding protein belonging to the apolipoprotein gene family. The protein is thought to play a role in lipid metabolism. Polymorphisms in this gene may influence circulating lipid levels and may be associated with coronary artery disease risk. This gene is present in a cluster with other related apolipoprotein genes on chromosome 19. Naturally occurring read-through transcription exists between this gene and the neighboring downstream apolipoprotein C-II (APOC2) gene.

Type: Primary
Antigen: APOC4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
PublicMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR