Anti-MAPK8IP1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-6255R-CY5
BOSSBS-6255R-CY5EA 843 GBP
BOSSBS-6255R-CY5
Anti-MAPK8IP1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
The JNK-interacting protein (JIP) group of scaffold proteins selectively mediates JNK signaling by aggregating specific components of the MAPK cascade to form a functional JNK signaling module. Required for JNK activation in response to excitotoxic stress. Cytoplasmic MAPK8IP1 causes inhibition of JNK-regulated activity by retaining JNK in the cytoplasm and inhibiting JNK phosphorylation of c-Jun. May also participate in ApoER2-specific reelin signaling. Directly, or indirectly, regulates GLUT2 gene expression and beta-cell function. Appears to have a role in cell signaling in mature andMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol MAPK8IP1
Antigen name Mitogen-Activated Protein Kinase 8 Interacting Protein 1
Environmentally Preferable
Antigen synonyms islet-brain 1|JNK-interacting protein 1|IB1|IB-1|JIP1|C-Jun-amino-terminal kinase-interacting protein 1|JNK MAP kinase scaffold protein 1|JIP-1|PRKM8 interacting protein|PRKM8IP
Antibody type Primary
Clonality Polyclonal
Conjugation Cy5®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 9479
Isotype IgG
ImmunoChemistry Yes
Immunogen 435-485/711
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR