Anti-LETMD1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
BS-5937R-A680
BOSSBS-5937R-A680EA 834 GBP
BOSSBS-5937R-A680
Anti-LETMD1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
This gene encodes a mitochondrial outer membrane protein. It has a potential role in tumorigenesis, which may result from negative regulation of the p53 tumor suppressor gene. Alternatively spliced transcript variants have been noted for this gene.

Type: Primary
Antigen: LETMD1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 21-120/360
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR