Anti-FGF6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Supplier: Bioss
BS-5923R-A750
BOSSBS-5923R-A750EA 834 GBP
BOSSBS-5923R-A750
Anti-FGF6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
FGF6 is a secreted heparin binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF6 is expressed in leukemia cell lines with platelet megakaryocytic differentiation potential. It signals through FGFR 1c, 2c, and 4. The FGF6 gene displays oncogenic transforming activity when transfected into mammalian cells. The mouse homolog of this gene exhibits a restricted expression profile predominantly inMore Product Information