Anti-CTAG1B Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-4503R-CY5.5
BOSSBS-4503R-CY5.5EA 843 GBP
BOSSBS-4503R-CY5.5
Anti-CTAG1B Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
The protein encoded by this gene is an antigen that is overexpressed in many cancers but that is also expressed in normal testis. This gene is found in a duplicated region of the X-chromosome and therefore has a neighboring gene of identical sequence. [provided by RefSeq, Jan 2012]

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CTAG1B
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR