Anti-TPX2 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-4285R
BOSSBS-4285REA 679 GBP
BOSSBS-4285R
Anti-TPX2 Rabbit Polyclonal Antibody
Antibodies
TPX2 is essential for spindle pole and mitotic spindle formulation, playing a role in the S and G2 phases of mitosis. It is also critical for the function of protein kinase Aurora A. TPX2 autophosphorylates Aurora A, targeting it to the spindle apparatus and allowing it to function in eukaryotic meiotic and mitotic cell cycles. TPX2 has also been found to be highly expressed in cancer cells.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: TPX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol TPX2
Antigen name TPX2, Microtubule Nucleation Factor
Environmentally Preferable
Antigen synonyms DIL-2|Targeting protein for Xklp2|C20orf1|Protein FLS353|DIL2|Hepatocellular carcinoma-associated antigen 519|C20orf2|p100|Restricted expression proliferation associated protein 100|Differentially expressed in cancerous and noncancerous lung cells 2|TPX2|HCA519
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 22974
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A.
Concentration 1 ug/ul
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR