Anti-SLC25A11 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-4037R-A555
BOSSBS-4037R-A555EA 837 GBP
BOSSBS-4037R-A555
Anti-SLC25A11 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
SLC25A11 catalyzes the transport of 2-oxoglutarate across the inner mitochondrial membrane in an electroneutral exchange for malate or other dicarboxylic acids, and plays an important role in several metabolic processes, including the malate-aspartate shuttle, the oxoglutarate/isocitrate shuttle, in gluconeogenesis from lactate, and in nitrogen metabolism.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC25A11
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR