Anti-SLC25A10 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BS-3997R-HRP
BOSSBS-3997R-HRPEA 843 GBP
BOSSBS-3997R-HRP
Anti-SLC25A10 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Involved in translocation of malonate, malate and succinate in exchange for phosphate, sulfate, sulfite or thiosulfate across mitochondrial inner membrane.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: SLC25A10
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR