Anti-T1 gamma Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
BS-3582R-A680
BOSSBS-3582R-A680EA 834 GBP
BOSSBS-3582R-A680
Anti-T1 gamma Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
Acts as an E3 ubiquitin-protein ligase. Promotes SMAD4 ubiquitination, nuclear exclusion and degradation via the ubiquitin proteasome pathway. According to PubMed:16751102, does not promote a decrease in the level of endogenous SMAD4. May act as a transcriptional repressor. Inhibits the transcriptional response to TGF-beta/BMP signaling cascade. Plays a role in the control of cell proliferation. Its association with SMAD2 and SMAD3 stimulates erythroid differentiation of hematopoietic stem/progenitor (By similarity). Monoubiquitinates SMAD4 and acts as an inhibitor of SMAD4-dependent TGF-More Product Information
Order Now

SPECIFICATIONS

Antigen name T1 gamma
Environmentally Preferable
Antigen synonyms RET-fused gene 7 protein|Tripartite motif-containing protein 33|TIF1GAMMA|KIAA1113|TIFGAMMA|TF1G|E3 ubiquitin-protein ligase TRIM33|Protein Rfg7|TIF1G|Transcription intermediary factor 1-gamma|RFG7|TIF1-gamma|PTC7|Ectodermin homolog|ECTO|TRIM33
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 680
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 51592
Isotype IgG
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Modification Unmodified
Epitope 981-1127/1127
Immunogen KLH conjugated synthetic peptide derived from human TIF1 gamma
Form Liquid
Molecular weight 124 kDa
Purification Purified by Protein A
Concentration 1 µg/µl
Storage buffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Storage temperature Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR