Anti-EIF2AK2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-3337R-A555
BOSSBS-3337R-A555EA 863 GBP
BOSSBS-3337R-A555
Anti-EIF2AK2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
IFN-induced dsRNA-dependent serine/threonine-protein kinase which plays a key role in the innate immune response to viral infection and is also involved in the regulation of signal transduction, apoptosis, cell proliferation and differentiation. Exerts its antiviral activity on a wide range of DNA and RNA viruses including hepatitis C virus (HCV), hepatitis B virus (HBV), measles virus (MV) and herpes simplex virus 1 (HHV-1). Inhibits viral replication via phosphorylation of the alpha subunit of eukaryotic initiation factor 2 (EIF2S1), this phosphorylation impairs the recycling of EIF2S1More Product Information
Order Now

SPECIFICATIONS

Antigen symbol EIF2AK2
Antigen name eukaryotic translation initiation factor 2-alpha kinase 2
Environmentally Preferable
Antigen synonyms double-stranded RNA-activated protein kinase|PRKR|Interferon-induced|EIF2AK1|eIF-2A protein kinase 2|Eukaryotic translation initiation factor 2-alpha kinase 2|PKR
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 555
Reactivity Human
Host Rabbit
Gene ID 5610
Isotype IgG
ImmunoChemistry Yes
Modification phospho-Thr446/Thr451
Immunogen 420-470/551
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR