Anti-NPM Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-3309R-CY7
BOSSBS-3309R-CY7EA 870 GBP
BOSSBS-3309R-CY7
Anti-NPM Rabbit Polyclonal Antibody (Cy7®)
Antibodies
This gene encodes a phosphoprotein which moves between the nucleus and the cytoplasm. The gene product is thought to be involved in several processes including regulation of the ARF/p53 pathway. A number of genes are fusion partners have been characterized, in particular the anaplastic lymphoma kinase gene on chromosome 2. Mutations in this gene are associated with acute myeloid leukemia. More than a dozen pseudogenes of this gene have been identified. Alternative splicing results in multiple transcript variants.[provided by RefSeq, Nov 2009].

Recommended Dilutions: IF(IHC-P): 1:50-200

Order Now

SPECIFICATIONS

Antigen symbol NPM
Environmentally Preferable
Antigen name Nucleophosmin
Antigen synonyms MGC104254|NO38|Nucleophosmin|Nucleolar protein NO38|numatrin|B23|Nucleolar phosphoprotein B23|NPM1
Antibody type Primary
Clonality Polyclonal
Conjugation Cy7®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 4869
Isotype IgG
ImmunoChemistry Yes
Modification phospho-Thr95
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR