Anti-Nuclear Matrix Protein p84/THOC1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-2448R-A488
BOSSBS-2448R-A488EA 837 GBP
BOSSBS-2448R-A488
Anti-Nuclear Matrix Protein p84/THOC1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
This protein is part of the THO complex (THOC). The THO complex is recruited to transcribed genes and travels with the RNA polymerase during elongation. It may physically link proteins that function in transcription and in RNA export and RNA processing.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Nuclear Matrix Protein p84/THOC1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen name Nuclear Matrix Protein p84/THOC1
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 488
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 9984
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR