Anti-OLR1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-2044R-A555
BOSSBS-2044R-A555EA 844 GBP
BOSSBS-2044R-A555
Anti-OLR1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Receptor that mediates the recognition, internalization and degradation of oxidatively modified low density lipoprotein (oxLDL) by vascular endothelial cells. OxLDL is a marker of atherosclerosis that induces vascular endothelial cell activation and dysfunction, resulting in pro-inflammatory responses, pro-oxidative conditions and apoptosis. Its association with oxLDL induces the activation of NF-kappa-B through an increased production of intracellular reactive oxygen and a variety of pro-atherogenic cellular responses including a reduction of nitric oxide (NO) release, monocyte adhesion andMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol OLR1
Antigen name Oxidized Low Density Lipoprotein Receptor 1
Environmentally Preferable
Antigen synonyms SCARE1|CLEC8A|LOX1|SLOX1|LOXIN
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 555
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 100009322
Isotype IgG
ImmunoChemistry Yes
Immunogen 224-274/274
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR