Anti-F2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Supplier: Bioss
BS-1914R-A350
BOSSBS-1914R-A350EA 837 GBP
BOSSBS-1914R-A350
Anti-F2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Thrombin is the final protease in the blood coagulation cascade and serves both pro- and anticoagulant functions through the cleavage of several targets. The ability of thrombin to specifically recognize a wide range of substrates derives from interactions which occur outside of the active site of thrombin. Thrombin possesses two anion binding exosites which mediate many of its interactions with cofactors and substrates, and although many structures of thrombin have been solved, few such interactions have been described in molecular detail. Glycosaminoglycan binding to exosite II of thrombinMore Product Information