Anti-HPV16 E6 + HPV18 E6 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss
BS-1719R-FITC
BOSSBS-1719R-FITCEA 843 GBP
BOSSBS-1719R-FITC
Anti-HPV16 E6 + HPV18 E6 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Transcriptional transactivator. Binds double-stranded DNA (By similarity). Has transforming activity. Inactivates, with E6-AP ubiquitin-protein ligase, the human p53/TP53 tumor suppressor protein by targeting it to degradation. Binds and targets human MUPP1/MPDZ protein to degradation. Those two functions presumably contribute to transforming activity. Interaction with human FBLN1 protein also seems to be linked to cell transformation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HPV16 E6 + HPV18 E6
Clonality: Polyclonal
Clone:
Conjugation: FITC (FluoresceinMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol HPV16 E6 + HPV18 E6
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Virus
Host Rabbit
Gene ID 1489088
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR