Anti-THOC2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss
BS-1663R-FITC
BOSSBS-1663R-FITCEA 843 GBP
BOSSBS-1663R-FITC
Anti-THOC2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Required for efficient export of polyadenylated RNA and spliced mRNA. Acts as component of the THO subcomplex of the TREX complex which is thought to couple mRNA transcription, processing and nuclear export, and which specifically associates with spliced mRNA and not with unspliced pre-mRNA. TREX is recruited to spliced mRNAs by a transcription-independent mechanism, binds to mRNA upstream of the exon-junction complex (EJC) and is recruited in a splicing- and cap-dependent manner to a region near the 5' end of the mRNA where it functions in mRNA export to the cytoplasm via the TAP/NFX1More Product Information
Order Now

SPECIFICATIONS

Antigen symbol THOC2
Antigen name THO Complex 2
Environmentally Preferable
Antigen synonyms hTREX120|THO2|CXorf3|dJ506G2.1|THOC2|THO complex subunit 2
Antibody type Primary
Clonality Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 57187
Isotype IgG
ImmunoChemistry Yes
Immunogen 130-180/1593
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR