Anti-TPA Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-1545R-CY3
BOSSBS-1545R-CY3EA 852 GBP
BOSSBS-1545R-CY3
Anti-TPA Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Converts the abundant, but inactive, zymogen plasminogen to plasmin by hydrolyzing a single Arg-Val bond in plasminogen. By controlling plasmin-mediated proteolysis, it plays an important role in tissue remodelling and degradation, in cell migration and many other physiopathological events. Plays a direct role in facilitating neuronal migration.

Type: Primary
Antigen: TPA
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 485-562/562
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR