Anti-DDIT3 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-1361R
BOSSBS-1361REA 705 GBP
BOSSBS-1361R
Anti-DDIT3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DDIT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR