Anti-GOLGA3 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-13480R-CY7
BOSSBS-13480R-CY7EA 843 GBP
BOSSBS-13480R-CY7
Anti-GOLGA3 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
The Golgi apparatus consists of a series of stacked, flattened membrane stacks called cisternae that are involved in the transport of lipids and proteins in the secretory pathway and are important for Golgi-microtubule interaction. Golgin 160 is a 1,498 amino acid protein that localizes to both the cytoplasm and to the Golgi apparatus and contains a series of coiled-coil domains. Expressed in a variety of tissues, including heart, liver, testis, kidney, lung and salivary gland, golgin 160 functions as a homodimer that interacts with GOLGA7 and is thought to be involved in maintaining GolgiMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol GOLGA3
Environmentally Preferable
Antigen name Golgin Subfamily A Member 3
Antigen synonyms golgin-165|Golgi complex-associated protein of 170 kD|SY2/SY10 protein|Golgi peripheral membrane protein|HGNC:4426|golgin subfamily a|Golgi autoantigen|MEA-2 |male enhanced antigen-2|golgin-160|Golgi membrane associated protein|GOLGA3|3|GCP170
Antibody type Primary
Clonality Polyclonal
Conjugation Cy7®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 2802
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR