Anti-GALNT1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BS-13273R-A647
BOSSBS-13273R-A647EA 837 GBP
BOSSBS-13273R-A647
Anti-GALNT1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
The UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAc-T) family of enzymes are substrate-specific proteins that catalyze the transfer of GalNAc (N-acetylgalactosamine) to serine and threonine residues onto various proteins, thereby initiating mucin-type O-linked glycosylation in the Golgi apparatus. GalNAc-T1, also known as GALNT1 (Polypeptide N-acetylgalactosaminyltransferase 1), is a ubiquitously expressed 559 amino acid single-pass type II membrane protein that localizes to the Golgi apparatus and, like other GalNAc-Ts, contains a stem region and a C-More Product Information
Order Now

SPECIFICATIONS

Antigen symbol GALNT1
Environmentally Preferable
Antigen name Polypeptide N-Acetylgalactosaminyltransferase 1
Antigen synonyms polypeptide N-acetylgalactosaminyltransferase 1|GALNAC-T1
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 647
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 2589
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR