Anti-ETV6 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-13115R-A488
BOSSBS-13115R-A488EA 837 GBP
BOSSBS-13115R-A488
Anti-ETV6 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Transcriptional repressor; binds to the DNA sequence 5'-CCGGAAGT-3'. Plays a role in hematopoiesis and malignant transformation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ETV6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

SPECIFICATIONS

Antigen symbol ETV6
Antigen name ETS variant 6
Environmentally Preferable
Antigen synonyms TEL/ABL|ETV6|ETS translocation variant 6|THC5|Transcription factor ETV6|TEL|TEL1|ETS-related protein Tel1
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 488
Reactivity Human,Mouse
Host Rabbit
Gene ID 2120
Isotype IgG
ImmunoChemistry Yes
Immunogen 80-130/452
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR