Anti-DCD Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-12996R-A488
BOSSBS-12996R-A488EA 837 GBP
BOSSBS-12996R-A488
Anti-DCD Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Antimicrobial peptides participate in the innate response, which may provide a barrier for protection against infection. The Dermcidin gene encodes an antimicrobial peptide DCD-1, which is constitutively expressed in sweat glands, secreted into the sweat, and transported to the epidermal surface. DCD-1 displays antimicrobial activity in response to a variety of pathogenic microorganisms. Overexpression of Dermcidin in breast cancers promotes cell growth and survival, and is coupled with a focal copy number gain of its locus on human chromosome 12q13.2.

Recommended Dilutions: IF(IHC-P): 1:5More Product Information
Order Now

SPECIFICATIONS

Antigen symbol DCD
Environmentally Preferable
Antigen name Dermcidin
Antigen synonyms Survival-promoting peptide|AIDD|DSEP|Dermcidin|DCD-1|DCD|Preproteolysin
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 488
Reactivity Human
Host Rabbit
Gene ID 117159
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR