Anti-KIF13B Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Supplier: Bioss
BS-12387R-A680
BOSSBS-12387R-A680EA 834 GBP
BOSSBS-12387R-A680
Anti-KIF13B Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
KIF13B is also known as Kinesin-like protein GAKIN or GAKIN and is a 1826 amino acid protein that is widely expressed in tissues, with highest expression in brain and kidney. KIF13B is localised to the cytoplasm, as well as to the cytoskeleton, and is thought to be a microtubule-dependent motor protein which is able to bind to a variety of proteins in order to traffic them to various locations throughout the cell. KIF13B belongs to the kinesin-like protein family and possesses three domains typical of the kinesin-like protein family, namely an N-terminal motor domain with an ATP-bindingMore Product Information