Anti-Synaptogyrin 4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Supplier: Bioss
BS-11375R-A750
BOSSBS-11375R-A750EA 834 GBP
BOSSBS-11375R-A750
Anti-Synaptogyrin 4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
The synaptogyrin family of proteins are integral membrane proteins containing four transmembrane regions. Synaptogyrins are tyrosine-phosphorylated proteins with two neuronal (Synaptogyrins 1 and 3) and one ubiquitous (Synaptogyrin-2) isoform. Synaptophysin and synaptogyrin represent the major constituents of synaptic vesicles. The 26 kDa protein Synaptogyrin-1 is associated with presynaptic vesicles in neuronal cells. Synaptogyrin-2, also known as Cellugyrin has a tyrosine phosphorylated C-terminal cytoplasmic tail, and is involved in the regulation of membrane traffic in non-neuronal cells.More Product Information