Anti-LMNB2 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-11132R-CY5
BOSSBS-11132R-CY5EA 837 GBP
BOSSBS-11132R-CY5
Anti-LMNB2 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
A unique family of Cysteine proteases has been described that differs in sequence, structure and substrate specificity from any previously described protease family. This family, termed CED-3/ICE, functions as key components of the apoptotic machinery and act to destroy specific target proteins which are critical to cellular longevity. Nuclear lamins are critical to maintaining the integrity of the nuclear envelope and cellular morphology as components of the nuclear lamina, a fibrous layer on the nucleoplasmic side of the inner nuclear membrane, which is thought to provide a framework forMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol LMNB2
Antigen name Lamin B2
Antigen synonyms LMN B2|LMN2|LMNB2|LMNB 2|LAMB2|MGC2721.|LMN 2|LMNB2_HUMAN|LAMB 2|Lamin-B2
Antibody type Primary
Clonality Polyclonal
Conjugation Cy5®
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR