Anti-HA Tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: Bioss
BS-0966R-HRP
BOSSBS-0966R-HRPEA 341 GBP
BOSSBS-0966R-HRP
Anti-HA Tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Human influenza hemagglutinin (HA) is a surface glycoprotein required for the infectivity of the human virus. The HA tag is derived from the HA molecule corresponding to amino acids 98-106 has been extensively used as a general epitope tag in expression vectors. Many recombinant proteins have been engineered to express the HA tag, which does not appear to interfere with the bioactivity or the biodistribution of the recombinant protein. This tag facilitates the detection, isolation, and purification of the proteins.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500