Anti-SLC33A1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-0699R-A350
BOSSBS-0699R-A350EA 837 GBP
BOSSBS-0699R-A350
Anti-SLC33A1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Probable acetyl-CoA transporter necessary for O-acetylation of gangliosides (PubMed:9096318). Negatively regulates BMP signaling (PubMed:25402622).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC33A1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR