Screw caps with aperture, BOLA
Supplier: Bohlender
H989-40
BOHLH989-40EA 9.3 GBP
BOHLH989-40
Screw caps with aperture, BOLA
Closures Bottle Closures
Screw cap, made of PPS, with handy knurl, with aperture.
  • Diameter of aperture: 28 mm
  • Universal chemical resistance
  • Temperature resistance: from –20 to +250 °C
  • Suitable gasket rings are available separately
As joining piece between tubes with flange and tubes with GL thread.