Snap valves, BOLA
Supplier: Bohlender
E674-54 E674-56
BOHLE674-54EA 622 GBP
BOHLE674-54 BOHLE674-56
Snap valves, BOLA
Stopcocks
Body made of PTFE with 2 connections. Nuts made of glass-fibre reinforced PTFE for connecting tubing or tube.
  • Motionless sealing by means of a gastight bellow made of PTFE with flat cone point, no leakages
  • Temperature resistant upto 110 °C
  • Universal chemical resistance
  • Pressure upto 6 bar and suitable for vacuum
Snap valves suitable for panel mounting and easy disassembly for cleaning.
Lever locks when valve is open, switch setting is easily visible at position of the lever.