Bossheads, three angles
Supplier: BOCHEM
5655 5656
BOCH5655EA 24.1 GBP
BOCH5655 BOCH5656
Bossheads, three angles
Clamps Bossheads
Three angled bossheads made of duraluminium (aluminium alloy).
  • Angle: 90 + 180 + 270°
  • Suitable for multilevel and three dimension constructions