Anti-Ascorbate Oxidase Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB20746-1
BIRBORB20746-1EA 0 GBP
BIRBORB20746-1
Anti-Ascorbate Oxidase Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Ascorbate Oxidase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Plant
Order Now

SPECIFICATIONS

Antigen name Ascorbate Oxidase
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Plant
Host Rabbit
Isotype
Immunogen Ascorbate oxidase isolated and purified from cucurbita species. Freunds complete adjuvant is used in the first step of the immunization procedure.
Form lyophilized
Concentration 1 mg/0,5ml, no foreign proteins added
Storage buffer PBS
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR