Models of brains
Supplier: 3B Scientific
1008552 1001261 1001262
BINH1008552EA 599 GBP
BINH1008552 BINH1001261 BINH1001262
Models of brains
Models Human Biology Models
These human brains are a very useful teaching aid, especially for large groups of students.
  • Unbreakable durable vinyl, can be broken down
  • Structures of the brain and ventricles are visible
  • Median, frontal, and transverse divisions
Delivery information: Delivered on a removable base and with a detailed product manual.
Order Now
SPECIFICATIONS
Environmentally Preferable