Environmental test chambers, MKF series
Supplier: Binder
9020-0380 9020-0378 9020-0379 9020-0381 9020-0408 9020-0409
BIND9020-0380EA 0 GBP
BIND9020-0380 BIND9020-0378 BIND9020-0379 BIND9020-0381 390-1669 390-1670
Environmental test chambers, MKF series
Testing Chambers Environmental Test Chambers
The MKF series of dynamic climate chambers has been specially developed as precision heating and cooling chambers for industrial material testing and environment simulation applications. The units are ideal for rapid temperature changes with humidity control and have a higher capacity which exceeds the capability of normal testing chambers. The stainless steel inner chamber has door heating, a heated viewing window with LED interior lighting and programmable condensation protection for the test materials.
  • MCS controller can activate four zero-voltage relay contacts
  • APT line™ preheating
More Product Information