Sedimentation cones, Imhoff type
Supplier: SciLabware
CRF530
BIBBCRF530EA 410 GBP
BIBBCRF530
Sedimentation cones, Imhoff type
Funnels Sedimentation Cones
Borosilicate glass.
  • Graduated up to 40 ml and with capacity mark at 1000 ml
For determining the amount of sediment in sewage or industrial waste.