Cleanroom wipes, Durx® 770
Supplier: BERKSHIRE
DR770090920 DR770.1818.10 DR770.0404.10 DR770121220
115-0345EA 126 GBP
115-0345 BERKDR770.1818.10 115-1785 BERKDR770121220
Cleanroom wipes, Durx® 770
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These wipes are composed of a hydroentangled non woven blend of 55% cellulose and 45% polyester. This combination of properties provides Durx® 770 with the level of absorbency, abrasion resistance and chemical compatibility required for controlled environments and applications.
  • Enhanced with an ultra low particle and fiber treatment
  • Excellent combination of the synthetic polyester strength and cleanliness with the absorbent characteristics of cellulose
  • Reduced particle and fiber generation with ultra low particle and fiber treatment
  • Smooth, absorbent and
More Product Information