Tissue culture tubes, Leighton type
Supplier: BELLCO GLASS
1908-16125
BELC1908-16125EA 775 GBP
BELC1908-16125
Tissue culture tubes, Leighton type
Tubes Culture Tubes
Glass, clear.
Leighton culture tubes are useful for the study of pathogenic effects on viruses on cells as well as sponge matrix culture and nutritional studies. Use cover slips for preparing explant tissue cultures with Leighton culture tubes.