Tanks, rectangular, heavy duty, with top flanges, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
H340850000 H340940000 H340950000 H340930000 H340920000
BELAH340850000EA 577 GBP
BELAH340850000 BELAH340940000 BELAH340950000 BELAH340930000 BELAH340920000
Tanks, rectangular, heavy duty, with top flanges, SP Bel-Art
Tanks Rectangular Tanks
These seamless, rotational-moulded PE tanks are lightweight and easy-to-handle.
  • Available with or without a faucet
  • Acid and alkali resistant
  • With wide top flanges, include covers
Ordering information: Can be supplied with spigots, fittings, dams, partitions etc. Contact local Avantor sales office for quotations on special sizes and fabrication in PP and PVC.
Order Now
SPECIFICATIONS
Environmentally Preferable