Petri dish rack, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F189850100
BELAF189850100EA 384 GBP
BELAF189850100
Petri dish rack, SP Bel-Art
Racks Petri Dish Racks
Clear TPX body, PP post. This Petri dish rack holds up to 54 (Ø 100 mm) dishes.
  • Finger slots for easy removal of dishes
  • Built-in handle for easy transportation
  • Rubber feet prevent rack from sliding on the benchtop
W×L×H: 200×340×230 mm.