Racks for centrifuge tubes, floating, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F188450080
BELAF188450080EA 91.4 GBP
BELAF188450080
Racks for centrifuge tubes, floating, SP Bel-Art
Racks Tube Racks
PP, white.
  • Durable and corrosion resistant
  • Easy transport and stacking
  • Work on the benchtop, float in the water bath
This open style floating rack provides tubes with maximum contact with the water and better circulation of the water bath around the tubes, allowing superior temperature control of samples.