Experimentation kits, conservation of genetic information
Supplier: G-Biosciences
BE-409
BE-409EA 360 GBP
BE-409
Experimentation kits, conservation of genetic information
Educational Materials Biology Educational Materials Biochemistry Learning Activities
In this lab activity, students learn to perform carefully controlled experiments to generate protein fragments using a proteolysis enzyme and then analyse the protein fragments by electrophoresis. By analysis of protein fragmentation patterns, i.e. protein fingerprints, students learn about protein sequence, structure, and their conservation. Students resolve a set of three functionally identical protein samples selected from throughout the animal kingdom; including human, bovine and sheep. After generating fingerprints by electrophoresis, students examine the protein fingerprint of eachMore Product Information